ContactUsLinkComponent
Cartridge, SPE, Normal Phase, Material: Polypropylene tubes with polyethylene frits, Silica (EPA SW 846 methods), Meets requirements of EPA Methods 515.2 and 553, Hydrophilic (polar) adsorbents used to extract hydrophilic analytes from nonpolar matrices, Volume: 6 mL, 1,000 mg
Catalog #: 76458-096
Supplier:  Restek
Cartridge, SPE, Normal Phase, Material: Polypropylene tubes with polyethylene frits, Silica (EPA SW 846 methods), Meets requirements of EPA Methods 515.2 and 553, Hydrophilic (polar) adsorbents used to extract hydrophilic analytes from nonpolar matrices, Volume: 6 mL, 1,000 mg
Catalog #: 76458-096
Supplier:  Restek
Customers Who Bought This Also Bought