VWR® Disposable Scalpels with Measurements
Catalog #: 76457-488
Supplier: VWR International
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Blade No.:No. 12
- Description:Disposable scalpel, sterile
- Cat. no.:76457-488
Specifications
About this item
VWR® Sterile Disposable Scalpels are mounted on plastic handles with fluted finger-grips.
- Wrapped in individual peel pouch
- Sterilized by gamma radiation
- Preassembled - Mounted on plastic handles with fluted finger-grips
Sterile. 10 per pack.
Certifications: ISO, GMP
Customers Who Bought This Also Bought