Wipe, Pre-Saturated Polypropylene, Colour: Blue, Sterile, Validated to SAL (Sterility Assurance Level) 10-6 in accordance with AAMI/ISO 11137:2006 guidelines, Cobalt-60 gamma irradiated, ISO Class 5, Meets and exceeds USP 797 and USP 800 requirements for critical contamination control, Size: 9x11in
Catalog #: 10210-951
Supplier: High-Tech Conversions
Customers Who Bought This Also Bought