ContactUsLinkComponent
Tris(hydroxymethyl)aminomethane, Biocertified, USP, EP, commonly used as a buffer and enzyme inhibitor in biopharmaceutical manufacturing, Synonyms: TRIS ; Trismat ; Trometamol ; Tromethamine, Storage: Hygroscopic, Keep tightly closed, Solid, Appearance: crystalline powder, Color: white, Size: 2500g
Catalog #: 76452-706
Supplier:  Spectrum Chemicals
Tris(hydroxymethyl)aminomethane, Biocertified, USP, EP, commonly used as a buffer and enzyme inhibitor in biopharmaceutical manufacturing, Synonyms: TRIS ; Trismat ; Trometamol ; Tromethamine, Storage: Hygroscopic, Keep tightly closed, Solid, Appearance: crystalline powder, Color: white, Size: 2500g
Catalog #: 76452-706
Supplier:  Spectrum Chemicals
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.