Earplug, Uncorded, No roll-down soft foam with dimpled tip collapses to be easily and gently pushed into ear canal, Bell Shape provides effective NRR 31dB, Canada Class A(L) hearing protection, Available in both uncorded and corded versions, stem fits naturally between the finger and thumb
Catalog #: 76452-384
Supplier: Honeywell Safety Products
Customers Who Bought This Also Bought