ContactUsLinkComponent
Lab-Based Mass Flow Calibrator, Whisper Series, 0-500SCCM, Monochrome Display, (requires power adaptor), Quickly and precisely verify flow rate generated by vacuum or pressure before going into the field, Ideal for real-time calibration of airflow controllers, Dimensions: 4.1in h x 2.4in w x 1.1in d
Catalog #: 76452-167
Supplier:  Restek
Lab-Based Mass Flow Calibrator, Whisper Series, 0-500SCCM, Monochrome Display, (requires power adaptor), Quickly and precisely verify flow rate generated by vacuum or pressure before going into the field, Ideal for real-time calibration of airflow controllers, Dimensions: 4.1in h x 2.4in w x 1.1in d
Catalog #: 76452-167
Supplier:  Restek
Customers Who Bought This Also Bought