VWR® Cleanroom Mask with Earloops, Made in USA
Catalog #: 76451-956
Supplier: VWR International
Specifications
- Sterility:Non sterile
- Color:White
- Description:VWR® cleanroom mask with earloops
- Cat. no.:76451-956
Specifications
About this item
The combination of strong BFE and PFE make these masks ideal for use in production or research areas.
- Protection against bacteria and particles
- 3-ply construction
- Non sterile
- With earloops
Part of the VWR® Made in USA PPE line, these cleanroom masks with earloops are a core item of any solid garment program, offering the adjustability user's desire.
Country of origin: USA.
Ordering information: Made in USA Masks with ties are also available; see catalog number 76409-318.
Packaging: Masks are packaged 50 per bag, 10 bags per case.
Customers Who Bought This Also Bought