ContactUsLinkComponent
Cannabigerolic Acid (CBGA) Standard, 1,000ug/ml, Acetonitrile, 1mL/ampul, Storage Temperature: -20 deg C or colder, Verified composition and stability, Prepared in acetonitrile, instead of methanol, to maintain stability and prevent methylation of carboxylic acid
Catalog #: 76452-150
Supplier:  Restek
Cannabigerolic Acid (CBGA) Standard, 1,000ug/ml, Acetonitrile, 1mL/ampul, Storage Temperature: -20 deg C or colder, Verified composition and stability, Prepared in acetonitrile, instead of methanol, to maintain stability and prevent methylation of carboxylic acid
Catalog #: 76452-150
Supplier:  Restek
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.