Vwr* Microbalance, U-Series, Readability: 1.0 ug, Plug Type: Switchable AC Plug (W2, W3U, W2E, W2A), Voltage: 100 - 240 V, Stabilization time: 8 s, Repeatabilty: 0.5 ug (at 5% load), platform size: 16 mm, Linearity: +/-3.0 ug, Temperature Range: 10-40 DEG C, Automatic level control, capacity: 2.1 g
Catalog #: MP76321-868
Supplier: VWR International
Customers Who Bought This Also Bought