Vwr* Ultra Microbalance, U-Series, Readability: 0.1 ug, Plug Type: Switchable AC Plug (W2, W3U, W2E, W2A), Voltage: 100 - 240 V, Stabilization time: 10 s, Repeatabilty: 0.25 ug (at 5% load), platform size: 16 mm, Linearity: +/-1.5 ug, Temp Range: 10-40DEG C, Automatic level control, capacity: 2.1 g
Catalog #: MP76321-866
Supplier: VWR International
Customers Who Bought This Also Bought