T-Piece Connection, for Multiple Units, Allows for the connection of two hoses onto one supply cylinder with constant flow through both openings, For use of filling two LN2 Auto-Fill tanks from one supply cylinder, Does not include valve
Catalog #: 76449-816
Supplier: Horizon Scientific
Customers Who Bought This Also Bought