Caps for Flowtubes for Flow Cytometry Instruments, Chemglass Life Sciences
Catalog #: 76449-662
Supplier: Chemglass
Specifications
- Sterility:Sterile
- Description:Strainer cap only for FlowTubes, with 35 µm strainer mesh
- Packaging:20 bags of 25