ContactUsLinkComponent
Centrifuge Tube, 30mL, Free Standing, 29 x 80mm, Polypropylene, Sterile, RNase, DNase, pyrogen and cytotoxin free, Compatible with standard rotors, Frosted marking surface, Tubes and caps may be frozen at temperatures as low as -86°C, centrifugal force is 7,000xg. Polyethylene caps, skirted bottom
Catalog #: 76449-656
Supplier:  Chemglass
Centrifuge Tube, 30mL, Free Standing, 29 x 80mm, Polypropylene, Sterile, RNase, DNase, pyrogen and cytotoxin free, Compatible with standard rotors, Frosted marking surface, Tubes and caps may be frozen at temperatures as low as -86°C, centrifugal force is 7,000xg. Polyethylene caps, skirted bottom
Catalog #: 76449-656
Supplier:  Chemglass
Customers Who Bought This Also Bought