ContactUsLinkComponent
Centrifuge Tube, 50mL, 29 x 115mm, Polypropylene, Individually Wrapped, Conical, Sterile, RNase, DNase, pyrogen and cytotoxin free, Compatible with standard rotors, Frosted marking surface, Tubes/caps may be frozen at temperatures as low as -86C, Caps made of polyethylene, Centrifugal force: 15000xg
Catalog #: 76449-654
Supplier:  Chemglass
Centrifuge Tube, 50mL, 29 x 115mm, Polypropylene, Individually Wrapped, Conical, Sterile, RNase, DNase, pyrogen and cytotoxin free, Compatible with standard rotors, Frosted marking surface, Tubes/caps may be frozen at temperatures as low as -86C, Caps made of polyethylene, Centrifugal force: 15000xg
Catalog #: 76449-654
Supplier:  Chemglass
Customers Who Bought This Also Bought