ContactUsLinkComponent
Centrifuge Tube, 15mL, 17 x 118mm, Polypropylene, Individually Wrapped, Sterile, RNase, DNase, pyrogen and cytotoxin free, Compatible with standard rotors, Frosted marking surface, Tubes and caps may be frozen at temperatures as low as -86°C, Caps are made of polyethylene, Centrifugal force: 12000xg
Catalog #: 76449-652
Supplier:  Chemglass
Centrifuge Tube, 15mL, 17 x 118mm, Polypropylene, Individually Wrapped, Sterile, RNase, DNase, pyrogen and cytotoxin free, Compatible with standard rotors, Frosted marking surface, Tubes and caps may be frozen at temperatures as low as -86°C, Caps are made of polyethylene, Centrifugal force: 12000xg
Catalog #: 76449-652
Supplier:  Chemglass
Customers Who Bought This Also Bought