ContactUsLinkComponent
Sample Tube, Centrifuge, 15mL, with a pre-attached 0.2um vent filter and 18in C-Flex Inlet, Polystyrene,(Purple cap), Pre-sterilized, Single use, Transfer of biopharmaceutical products and reagents in a closed system, Gas exchange transfer
Catalog #: 76449-640
Supplier:  Chemglass
Sample Tube, Centrifuge, 15mL, with a pre-attached 0.2um vent filter and 18in C-Flex Inlet, Polystyrene,(Purple cap), Pre-sterilized, Single use, Transfer of biopharmaceutical products and reagents in a closed system, Gas exchange transfer
Catalog #: 76449-640
Supplier:  Chemglass
Customers Who Bought This Also Bought