Empore™ Solid Phase Extraction Stage Tips, CDS Analytical
Catalog #: 76449-270
Supplier: CDS ANALYTICAL
Specifications
- Volume:200 µl
- Tip Style:6095, strong anion exchange
Specifications
About this item
The original Empore SPE membrane is now available in pre-packed StageTips and are the only solution to save time and improve reproducibility of peptide / protein desalting and fractionation.
- Save time by avoiding manual preparation of StageTips
- Reproducibility – Standardized packing methods to avoid tip-to-tip variability present in manual preparation in the lab
- Compatibility – Chemically resistant to organic solvents and acidic conditions and suitable for all protein and peptide desalting / fractionation applications
- Flexibility – Combine multiple phases into one tip or increase the number of layers for increased capacity
Empore membrane SPE StageTips are cost-effective and help users avoid manual preparation of StageTips while offering standardized packing methods to improve tip-to-tip consistency.
Particle size: 12 µm
Packaging: 96 tips per pack; 960 tips, or 10 packs, per case.
Customers Who Bought This Also Bought