Cylinder, To Deliver, Single Metric Scale, With Bumper Guard, Plastic Hexagonal Base, Made from borosilicate 3.3 Glass in heavy duty construction, reinforced top bead and with pour spout, Graduated Interval: 2 to 100ml, Sub Division: 1.0ml, Height: 248mm, Volume: 100ml
Catalog #: CA10823-007
Supplier: CHEM SCIENCE INC CA
Customers Who Bought This Also Bought