Cylinder, To Deliver, Single Metric Scale, With Bumper Guard, Plastic Hexagonal Base, Made from borosilicate 3.3 Glass in heavy duty construction, reinforced top bead and with pour spout, Graduated Interval: 2 to 50ml, Sub Division: 1.0ml, Height: 175mm, Volume: 50ml
Catalog #: CA10823-005
Supplier: CHEM SCIENCE INC CA
Customers Who Bought This Also Bought