Cylinder, To Deliver, Single Metric Scale, With Bumper Guard, Plastic Hexagonal Base, Made from borosilicate 3.3 Glass in heavy duty construction, reinforced top bead and with pour spout, Graduated Interval: 1 to 25ml, Sub Division: 0.5ml, Height: 138mm, Volume: 25ml
Catalog #: CA10823-003
Supplier: CHEM SCIENCE INC CA
Customers Who Bought This Also Bought