Cylinder, To Deliver, Single Metric Scale, With Bumper Guard, Plastic Hexagonal Base, Made from borosilicate 3.3 Glass in heavy duty construction, reinforced top bead and with pour spout, Graduated Interval: 0.4 to 20ml, Sub Division: 0.2ml, Height: 132mm, Volume: 10ml
Catalog #: CA10823-001
Supplier: CHEM SCIENCE INC CA
Customers Who Bought This Also Bought