ContactUsLinkComponent
Annexin V, Format: APC, Application: Flow Cytometry, belongs to the annexin family of calcium-dependent phospholipid binding proteins and has a high affinity for phosphatidylserine (PS), sensitive detection probe for early stage apoptotic change, Size: 100tests
Catalog #: 76449-054
Supplier:  Tonbo Biosciences
Annexin V, Format: APC, Application: Flow Cytometry, belongs to the annexin family of calcium-dependent phospholipid binding proteins and has a high affinity for phosphatidylserine (PS), sensitive detection probe for early stage apoptotic change, Size: 100tests
Catalog #: 76449-054
Supplier:  Tonbo Biosciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.