ContactUsLinkComponent
Kit, Annexin V Apoptosis Detection with 7-AAD, Format: APC, contains the reagents needed to identify apoptotic or necrotic cells by flow cytometry, Contains: APC Annexin V (100 tests), Annexin V Binding Buffer (1X) (50 mL), 7-AAD (200 tests)
Catalog #: 76449-050
Supplier:  Tonbo Biosciences
Kit, Annexin V Apoptosis Detection with 7-AAD, Format: APC, contains the reagents needed to identify apoptotic or necrotic cells by flow cytometry, Contains: APC Annexin V (100 tests), Annexin V Binding Buffer (1X) (50 mL), 7-AAD (200 tests)
Catalog #: 76449-050
Supplier:  Tonbo Biosciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.