ContactUsLinkComponent
CD274, Monoclonal antibody, Clone: 29E.2A3, Host: Mouse, Isotype: IgG2bk, Reactivity: Human, Format: APC, Application: Flow Cytometry, Synonyms: PD-L1, B7-H1, type 1 transmembrane glycoprotein and member of Ig superfamily, plays an important role in regulating immune cell responses, Size: 100tests
Catalog #: 76449-044
Supplier:  Tonbo Biosciences
CD274, Monoclonal antibody, Clone: 29E.2A3, Host: Mouse, Isotype: IgG2bk, Reactivity: Human, Format: APC, Application: Flow Cytometry, Synonyms: PD-L1, B7-H1, type 1 transmembrane glycoprotein and member of Ig superfamily, plays an important role in regulating immune cell responses, Size: 100tests
Catalog #: 76449-044
Supplier:  Tonbo Biosciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.