CD279, Monoclonal antibody, Clone: J110, Host: Mouse, Isotype: IgG1, Reactivity: Human, Format: APC, Synonyms: CD279, PD-1, PD1, SLEB2, hPD-1, hPD-l, hSLE1, Application: Flowcytometry, also known as (PD-1), 55 kDa, expressed on activated T cells, B cells, and myeloid cells, Size: 25 tests
Catalog #: 76449-038
Supplier: Tonbo Biosciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.