ContactUsLinkComponent
CD49d, Monoclonal antibody, Clone: 9F10, Host: Mouse, Isotype: IgG1, kappa, Reactivity: Human, Format: APC, Synonyms: VLA-4 chain, 4 integrin A, integrin A4 chain, ITGA4, Application: Flow cytometry, 150kD integral membrane protein that is also known as integrin alpha4, Size: 25 tests
Catalog #: 76449-026
Supplier:  Tonbo Biosciences
CD49d, Monoclonal antibody, Clone: 9F10, Host: Mouse, Isotype: IgG1, kappa, Reactivity: Human, Format: APC, Synonyms: VLA-4 chain, 4 integrin A, integrin A4 chain, ITGA4, Application: Flow cytometry, 150kD integral membrane protein that is also known as integrin alpha4, Size: 25 tests
Catalog #: 76449-026
Supplier:  Tonbo Biosciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.