ContactUsLinkComponent
CD80, Monoclonal antibody, Clone: 2D10.4, Host: Mouse, Isotype: IgG1, kappa, Reactivity: Human, Cross Reactivity: Rhesus, Format: APC, Application: Flow Cytometry, 60 kDa glycoprotein, member of Ig superfamily, also known as B7-1, is a ligand for the T cell receptors CD28 and CTLA-4, Size: 100 tests
Catalog #: 76449-024
Supplier:  Tonbo Biosciences
CD80, Monoclonal antibody, Clone: 2D10.4, Host: Mouse, Isotype: IgG1, kappa, Reactivity: Human, Cross Reactivity: Rhesus, Format: APC, Application: Flow Cytometry, 60 kDa glycoprotein, member of Ig superfamily, also known as B7-1, is a ligand for the T cell receptors CD28 and CTLA-4, Size: 100 tests
Catalog #: 76449-024
Supplier:  Tonbo Biosciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.