ContactUsLinkComponent
CD272, Monoclonal antibody, Clone: 6A6, Host: Armenian Hamster, Isotype: IgG, kappa, Format: APC, Application: Flowcytometry, type 1 transmembrane glycoprotein and member of the Ig superfamily, expressed on peripheral lymphocytes as well as developing B and T cells, macrophages, Size: 25 ug
Catalog #: 76449-002
Supplier:  Tonbo Biosciences
CD272, Monoclonal antibody, Clone: 6A6, Host: Armenian Hamster, Isotype: IgG, kappa, Format: APC, Application: Flowcytometry, type 1 transmembrane glycoprotein and member of the Ig superfamily, expressed on peripheral lymphocytes as well as developing B and T cells, macrophages, Size: 25 ug
Catalog #: 76449-002
Supplier:  Tonbo Biosciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.