CD40, Monoclonal antibody, Clone: FGK45, Host: Rat, Isotype: IgG2a, Reactivity: Mouse, Format: APC, Synonyms: Bp50, gp39, HIGM1, IGM, IMD3, T-BAM, TRAP, TNFRSF5, p50, FGK4.5, Application: Flowcytometry, 40-50 kDa glycoprotein member of the tumor necrosis factor receptor (TNFR) family, Size: 25 ug
Catalog #: 76448-986
Supplier: Tonbo Biosciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.