ContactUsLinkComponent
TIM-4, Monoclonal antibody, Clone: RMT4-54, Host: Rat, Isotype: IgG2a, K, Reactivity: Mouse, Format: APC, Synonyms: T cell immunoglobulin, Application: Flowcytometry, also known as T cell immunoglobulin and mucin domain containing protein-4, expressed on macrophages and dendritic cells, Size: 25 ug
Catalog #: 76448-966
Supplier:  Tonbo Biosciences
TIM-4, Monoclonal antibody, Clone: RMT4-54, Host: Rat, Isotype: IgG2a, K, Reactivity: Mouse, Format: APC, Synonyms: T cell immunoglobulin, Application: Flowcytometry, also known as T cell immunoglobulin and mucin domain containing protein-4, expressed on macrophages and dendritic cells, Size: 25 ug
Catalog #: 76448-966
Supplier:  Tonbo Biosciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.