ContactUsLinkComponent
Streptavidin, Monoclonal antibody, Format: APC, Synonyms: SAv-APC, SA-APC, Streptavidin-Allophycocyanin, Application: Flowcytometry, second-step reagent useful when detecting biotinylated antibodies in indirect staining protocols, bacterially derived, displays less nonspecific binding, Size: 500ug
Catalog #: 76448-960
Supplier:  Tonbo Biosciences
Streptavidin, Monoclonal antibody, Format: APC, Synonyms: SAv-APC, SA-APC, Streptavidin-Allophycocyanin, Application: Flowcytometry, second-step reagent useful when detecting biotinylated antibodies in indirect staining protocols, bacterially derived, displays less nonspecific binding, Size: 500ug
Catalog #: 76448-960
Supplier:  Tonbo Biosciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.