ContactUsLinkComponent
TER-119 Monoclonal antibody, Clone: TER-119, Host: Rat, Species reactivity: Rat, Isotype: IgG2b, Kappa, Format: APC-Cyanine7, Application: Flow Cytometry, TER-119 antigen is not expressed at either BFU-E or CFU-E stages, i.e. Prior to the pro-erythroblast stage, Size: 100 ug
Catalog #: 76448-944
Supplier:  Tonbo Biosciences
TER-119 Monoclonal antibody, Clone: TER-119, Host: Rat, Species reactivity: Rat, Isotype: IgG2b, Kappa, Format: APC-Cyanine7, Application: Flow Cytometry, TER-119 antigen is not expressed at either BFU-E or CFU-E stages, i.e. Prior to the pro-erythroblast stage, Size: 100 ug
Catalog #: 76448-944
Supplier:  Tonbo Biosciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.