ContactUsLinkComponent
CD4 Monoclonal antibody, Clone: RPA-T4, Host: Mouse, Species reactivity: Human, Isotype: IgG1, kappa, Format: Biotin, Application: Flow Cytometry, recognizes a different epitope, and thus does not block binding of, CD4 is typically expressed on thymocytes, some mature T cell populations, Size: 100ug
Catalog #: 76448-916
Supplier:  Tonbo Biosciences
CD4 Monoclonal antibody, Clone: RPA-T4, Host: Mouse, Species reactivity: Human, Isotype: IgG1, kappa, Format: Biotin, Application: Flow Cytometry, recognizes a different epitope, and thus does not block binding of, CD4 is typically expressed on thymocytes, some mature T cell populations, Size: 100ug
Catalog #: 76448-916
Supplier:  Tonbo Biosciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.