ContactUsLinkComponent
CD49b monoclonal Antibody, Clone: HMa2, Host: Armenian Hamster, Species reactivity: Mouse, Isotype: IgG, Format: Biotin, Applications: Flow Cytometryreacts with mouse CD49b, a 150 kDa glycoprotein member of the integrin family. CD49b, also known as integrin alpha2, size: 25ug
Catalog #: 76448-892
Supplier:  Tonbo Biosciences
CD49b monoclonal Antibody, Clone: HMa2, Host: Armenian Hamster, Species reactivity: Mouse, Isotype: IgG, Format: Biotin, Applications: Flow Cytometryreacts with mouse CD49b, a 150 kDa glycoprotein member of the integrin family. CD49b, also known as integrin alpha2, size: 25ug
Catalog #: 76448-892
Supplier:  Tonbo Biosciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.