ContactUsLinkComponent
CD62L monoclonal Antibody, Clone: MEL-14, Host: Rat, Species reactivity: Mouse, Isotype: 65-0032 , Format: Biotin, Synonyms: LECAM-1, Ly-22, Applications: Flow Cytometryfor mouse CD62L, also known as L-Selectin, cell adhesion molecule which facilitates lymphocyte rolling on endothelium, size: 25ug
Catalog #: 76448-888
Supplier:  Tonbo Biosciences
CD62L monoclonal Antibody, Clone: MEL-14, Host: Rat, Species reactivity: Mouse, Isotype: 65-0032 , Format: Biotin, Synonyms: LECAM-1, Ly-22, Applications: Flow Cytometryfor mouse CD62L, also known as L-Selectin, cell adhesion molecule which facilitates lymphocyte rolling on endothelium, size: 25ug
Catalog #: 76448-888
Supplier:  Tonbo Biosciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.