ContactUsLinkComponent
Annexin V Apoptosis Detection Kit, with 7-AAD, contains the reagents needed identify apoptotic or necrotic cells, Contains: FITC Annexin V 100tests, Annexin V Binding Buffer (1X) 50ml, 7-AAD 200tests, Format: FITC, Applications: Flow Cytometry
Catalog #: 76448-874
Supplier:  Tonbo Biosciences
Annexin V Apoptosis Detection Kit, with 7-AAD, contains the reagents needed identify apoptotic or necrotic cells, Contains: FITC Annexin V 100tests, Annexin V Binding Buffer (1X) 50ml, 7-AAD 200tests, Format: FITC, Applications: Flow Cytometry
Catalog #: 76448-874
Supplier:  Tonbo Biosciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.