ContactUsLinkComponent
Ghost Dye Blue 516, Excitation Laser: 488 nm, Emission: 516nm, Formulation: 1 uL/test in DMSO, Application: Flow Cytometry, amine reactive viability dye that can be used to discriminate viable from non-viable mammalian cells, irreversibly binds free amines available on cell surface, Size: 500tests
Catalog #: 76448-860
Supplier:  Tonbo Biosciences
Ghost Dye Blue 516, Excitation Laser: 488 nm, Emission: 516nm, Formulation: 1 uL/test in DMSO, Application: Flow Cytometry, amine reactive viability dye that can be used to discriminate viable from non-viable mammalian cells, irreversibly binds free amines available on cell surface, Size: 500tests
Catalog #: 76448-860
Supplier:  Tonbo Biosciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.