Annexin V, belongs to the annexin family of calcium-dependent phospholipid binding proteins and has a high affinity for phosphatidylserine, One of the characteristic features of early stage apoptosis is the translocation of PS, Format: PE, Applications: Flow Cytometry, size: 100 tests
Catalog #: 76448-826
Supplier: Tonbo Biosciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.