ContactUsLinkComponent
CD80 monoclonal Antibody, Clone: 2D10.4, Host: Mouse, Species reactivity: Human, Isotype: IgG1, Format: PE, Applications: Flow Cytometryreacts with human CD80, a 60 kDa glycoprotein that is a member of the Ig superfamILY, also known as B7-1, is ligand for the T cell receptors CD28, size: 25 tests
Catalog #: 76448-788
Supplier:  Tonbo Biosciences
CD80 monoclonal Antibody, Clone: 2D10.4, Host: Mouse, Species reactivity: Human, Isotype: IgG1, Format: PE, Applications: Flow Cytometryreacts with human CD80, a 60 kDa glycoprotein that is a member of the Ig superfamILY, also known as B7-1, is ligand for the T cell receptors CD28, size: 25 tests
Catalog #: 76448-788
Supplier:  Tonbo Biosciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.