ContactUsLinkComponent
CD366 monoclonal Antibody, Clone: RMT3-23, Host: Rat, Species reactivity: Mouse, Isotype: IgG2a, K, Format: PE, Synonyms: Havcr2, TIM3, Timd3, Applications: Flow Cytometryreacts with mouse CD366 which is also known as Tim-3 (T cell immunoglobulin and mucin domain containing protein-3), size: 100ug
Catalog #: 76448-760
Supplier:  Tonbo Biosciences
CD366 monoclonal Antibody, Clone: RMT3-23, Host: Rat, Species reactivity: Mouse, Isotype: IgG2a, K, Format: PE, Synonyms: Havcr2, TIM3, Timd3, Applications: Flow Cytometryreacts with mouse CD366 which is also known as Tim-3 (T cell immunoglobulin and mucin domain containing protein-3), size: 100ug
Catalog #: 76448-760
Supplier:  Tonbo Biosciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.