ContactUsLinkComponent
Isotype Control, Clone: HRPN, Host: Rat, Isotype: IgG1, HRPN immunoglobulin is useful as an isotype-matched control. The HRPN immunolglobulin has an unknown binding specificity and is used as an isotype control for IgG1 antibodies, Applications: Flow Cytometry, size: 100ug
Catalog #: 76448-720
Supplier:  Tonbo Biosciences
Isotype Control, Clone: HRPN, Host: Rat, Isotype: IgG1, HRPN immunoglobulin is useful as an isotype-matched control. The HRPN immunolglobulin has an unknown binding specificity and is used as an isotype control for IgG1 antibodies, Applications: Flow Cytometry, size: 100ug
Catalog #: 76448-720
Supplier:  Tonbo Biosciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.