ContactUsLinkComponent
CD279 monoclonal Antibody, Clone: RMP1-30, Host: Rat, Species reactivity: Mouse, Isotype: IgG2b, K, Format: PE-Cyanine7, Applications: Flow Cytometryfor mouse CD279, also known as programmed death-1 (PD-1), a 55 kDa glycoprotein member of the Ig superfamily of molecules, size: 25ug
Catalog #: 76448-698
Supplier:  Tonbo Biosciences
CD279 monoclonal Antibody, Clone: RMP1-30, Host: Rat, Species reactivity: Mouse, Isotype: IgG2b, K, Format: PE-Cyanine7, Applications: Flow Cytometryfor mouse CD279, also known as programmed death-1 (PD-1), a 55 kDa glycoprotein member of the Ig superfamily of molecules, size: 25ug
Catalog #: 76448-698
Supplier:  Tonbo Biosciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.