ContactUsLinkComponent
CD14 monoclonal Antibody, Clone: 61D3, Host: Mouse, Species reactivity: Human, Isotype: IgG1, Format: PerCP-Cyanine5.5, Synonyms: LPS-R, Mo2, Applications: Flow Cytometryfor human CD14, a 53-55 kDa GPI-anchored glycoprotein, highly expressed on monocytes and to a lesser extent, size: 100 tests
Catalog #: 76448-690
Supplier:  Tonbo Biosciences
CD14 monoclonal Antibody, Clone: 61D3, Host: Mouse, Species reactivity: Human, Isotype: IgG1, Format: PerCP-Cyanine5.5, Synonyms: LPS-R, Mo2, Applications: Flow Cytometryfor human CD14, a 53-55 kDa GPI-anchored glycoprotein, highly expressed on monocytes and to a lesser extent, size: 100 tests
Catalog #: 76448-690
Supplier:  Tonbo Biosciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.