ContactUsLinkComponent
CD252 monoclonal Antibody, Clone: RM134L, Host: Rat, Species reactivity: Mouse, Isotype: IgG2b, K, Format: PerCP-Cyanine5.5, Synonyms: CD134L, CD134 Ligand, OX-40L, OX-40 Ligand, TNF-ASF4, Tumor necrosis factor ligand superfamily member 4, gp34, Applications: Flow Cytometry, size: 100ug
Catalog #: 76448-666
Supplier:  Tonbo Biosciences
CD252 monoclonal Antibody, Clone: RM134L, Host: Rat, Species reactivity: Mouse, Isotype: IgG2b, K, Format: PerCP-Cyanine5.5, Synonyms: CD134L, CD134 Ligand, OX-40L, OX-40 Ligand, TNF-ASF4, Tumor necrosis factor ligand superfamily member 4, gp34, Applications: Flow Cytometry, size: 100ug
Catalog #: 76448-666
Supplier:  Tonbo Biosciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.