ContactUsLinkComponent
CD274 monoclonal Antibody, Clone: 29E.2A3, Host: Mouse, Species reactivity: Human, Isotype: IgG2b, Format: Purified, Synonyms: PD-L1, B7-H1, Applications: Flow Cytometryreacts with human CD274 which is also known as PD-L1 or B7-H1. CD274 is a type 1 transmembrane glycoprotein, size: 500ug
Catalog #: 76448-642
Supplier:  Tonbo Biosciences
CD274 monoclonal Antibody, Clone: 29E.2A3, Host: Mouse, Species reactivity: Human, Isotype: IgG2b, Format: Purified, Synonyms: PD-L1, B7-H1, Applications: Flow Cytometryreacts with human CD274 which is also known as PD-L1 or B7-H1. CD274 is a type 1 transmembrane glycoprotein, size: 500ug
Catalog #: 76448-642
Supplier:  Tonbo Biosciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.