ContactUsLinkComponent
CD154 monoclonal antibody, Clone: 5C8, Host: Mouse, Species reactivity: Human, Isotype: IgG2a, K, Format: Purified, Synonyms: CD40L, gp39, T-BAM, TRAP, Ly-62, Applications: Flow Cytometryreacts with human CD154 which is also known as CD40 ligand, 39kDa member of the TNF superfamily, size: 100ug
Catalog #: 76447-346
Supplier:  Tonbo Biosciences
CD154 monoclonal antibody, Clone: 5C8, Host: Mouse, Species reactivity: Human, Isotype: IgG2a, K, Format: Purified, Synonyms: CD40L, gp39, T-BAM, TRAP, Ly-62, Applications: Flow Cytometryreacts with human CD154 which is also known as CD40 ligand, 39kDa member of the TNF superfamily, size: 100ug
Catalog #: 76447-346
Supplier:  Tonbo Biosciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.