Isotype Control, Clone: 2A3, Host: Rat, Isotype: IgG2a, Format: violetFluor 500, Applications: Flow CytometryThe 2A3 immunoglobulin is useful as an isotype-matched control, binds to TNP (trinitrophenol), a hapten not expressed on mouse or human cells, size: 100ug
Catalog #: 76447-326
Supplier: Tonbo Biosciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.