ContactUsLinkComponent
Ly-6G monoclonal Antibody, Clone: 1A8, Host: Rat, Species reactivity: Mouse, Isotype: IgG2a, K, Format: violetFluor 500, Synonyms: Gr-1, Applications: Flow Cytometrybinds to mouse Ly-6G, commonly known as Gr-1, a member of the Ly-6 superfamily of GPI-anchored cell surface proteins, size: 100ug
Catalog #: 76447-294
Supplier:  Tonbo Biosciences
Ly-6G monoclonal Antibody, Clone: 1A8, Host: Rat, Species reactivity: Mouse, Isotype: IgG2a, K, Format: violetFluor 500, Synonyms: Gr-1, Applications: Flow Cytometrybinds to mouse Ly-6G, commonly known as Gr-1, a member of the Ly-6 superfamily of GPI-anchored cell surface proteins, size: 100ug
Catalog #: 76447-294
Supplier:  Tonbo Biosciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.