ContactUsLinkComponent
CD3 monoclonal Antibody, Clone: UCHT1, Host: Mouse, Species reactivity: Human, Isotype: IgG1, Format: violetFluor 500, Synonyms: Leu-4, T3, Applications: Flow Cytometryfor human CD3e, also known as CD3 epsilon, a 20 kDa subunit of the T cell receptor complex, size: 100 tests
Catalog #: 76447-274
Supplier:  Tonbo Biosciences
CD3 monoclonal Antibody, Clone: UCHT1, Host: Mouse, Species reactivity: Human, Isotype: IgG1, Format: violetFluor 500, Synonyms: Leu-4, T3, Applications: Flow Cytometryfor human CD3e, also known as CD3 epsilon, a 20 kDa subunit of the T cell receptor complex, size: 100 tests
Catalog #: 76447-274
Supplier:  Tonbo Biosciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.