ContactUsLinkComponent
Brefeldin A Solution (1000X), Brefeldin A (BFA) is a protein transport inhibitor that can be added to cell culture medium during in vitro activation of cells to inhibit the secretion of cytokines elicited by stimulation, Application: Functional Assays, Size: 1ml
Catalog #: 76447-262
Supplier:  Tonbo Biosciences
Brefeldin A Solution (1000X), Brefeldin A (BFA) is a protein transport inhibitor that can be added to cell culture medium during in vitro activation of cells to inhibit the secretion of cytokines elicited by stimulation, Application: Functional Assays, Size: 1ml
Catalog #: 76447-262
Supplier:  Tonbo Biosciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.